Gene Transcripts

Gene Symbol
Lrrc3c
Entrez Gene ID
287668
Gene Name
leucine rich repeat containing 3C
Gene Aliases
RGD1563281
Chromosome Location
Chr. 10: 86564080 - 86565695 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_220923.3 XP_220923.1 3 379

View Full Product Details