Gene Transcripts

Gene Symbol
Plcd3
Entrez Gene ID
287745
Gene Name
phospholipase C, delta 3
Gene Aliases
Chromosome Location
Chr. 10: 91164786 - 91186104 on Build Rnor_6.0
UniGene ID
Rn.95716
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105845.2 NP_001099315.2 9 1553
XM_006247494.2 XP_006247556.1 9 1594

View Full Product Details