Gene Transcripts

Gene Symbol
TNP1
Entrez Gene ID
7141
Gene Name
transition protein 1
Gene Aliases
TP1
Chromosome Location
Chr. 2: 216859458 - 216860059 on Build GRCh38
UniGene ID
Hs.3017
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003284.3 NP_003275.1 2 282
GenBank mRNA BC029516.1 - 2 286
X07948.1 - 2 314

View Full Product Details