Gene Transcripts

Gene Symbol
Ccl24
Entrez Gene ID
288593
Gene Name
chemokine (C-C motif) ligand 24
Gene Aliases
Chromosome Location
Chr. 12: 24148629 - 24152626 on Build Rnor_6.0
UniGene ID
Rn.138822
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013045.1 NP_001013063.1 3 273
GenBank mRNA BC087576.1 - 3 273

View Full Product Details