Gene Transcripts

Gene Symbol
TPTE
Entrez Gene ID
7179
Gene Name
transmembrane phosphatase with tensin homology
Gene Aliases
CT44, PTEN2
Chromosome Location
Chr. 21: 10521514 - 10606270 on Build GRCh38
UniGene ID
Hs.122986
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290224.1 NP_001277153.1 14, 15 1292
NM_199259.3 NP_954868.1 18, 19 1514
NM_199260.3 NP_954869.1 17, 18 1454
NM_199261.3 NP_954870.2 19, 20 1568
GenBank mRNA AF007118.1 - 19, 20 1516
AF495908.1 - 18, 19 1462
AK308254.1 - 18, 19 1411
AK314285.1 - 19, 20 1467
AL833345.1 - 19, 20 1475
AY219886.1 - 18, 19 1462
AY219887.1 - 17, 18 1402
AY219888.1 - 14, 15 1240
BC028719.1 - 18, 19 1497 5269598

View Full Product Details