Gene Transcripts

Gene Symbol
LOC289035
Entrez Gene ID
289035
Gene Name
similar to UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylgluco
Gene Aliases
Chromosome Location
Chr. 13: 51218472 - 51223840 on Build Rnor_6.0
UniGene ID
Rn.162322
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013872.1 NP_001013894.2 5 938
GenBank mRNA BC079259.1 - 5 938

View Full Product Details