Gene Transcripts
- Gene Symbol
- Sdhc
- Entrez Gene ID
- 289217
- Gene Name
- succinate dehydrogenase complex, subunit C, integral membrane protein
- Gene Aliases
- Chromosome Location
- Chr. 13: 89498047 - 89518939 on Build Rnor_6.0
- UniGene ID
- Rn.1698
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005534.1 |
NP_001005534.1
|
2, 3 |
85 |
|
XM_008769731.1 |
XP_008767953.1
|
3, 4 |
132 |
|
GenBank mRNA |
BC082027.1 |
- |
2, 3 |
85 |
|
FQ210665.1 |
- |
2, 3 |
87 |
|
FQ213406.1 |
- |
2, 3 |
104 |
|
FQ215668.1 |
- |
3, 4 |
139 |
|
FQ215689.1 |
- |
2, 3 |
87 |
|
FQ215712.1 |
- |
2, 3 |
87 |
|
FQ216122.1 |
- |
2, 3 |
103 |
|
FQ216612.1 |
- |
2, 3 |
104 |
|
FQ216658.1 |
- |
2, 3 |
87 |
|
FQ216892.1 |
- |
2, 3 |
103 |
|
FQ217679.1 |
- |
2, 3 |
104 |
|
FQ224704.1 |
- |
2, 3 |
87 |
|
FQ225195.1 |
- |
2, 3 |
98 |
|
FQ229651.1 |
- |
2, 3 |
104 |
|
FQ231805.1 |
- |
2, 3 |
104 |
|
View Full Product Details