Gene Transcripts

Gene Symbol
Corin
Entrez Gene ID
289596
Gene Name
corin, serine peptidase
Gene Aliases
Chromosome Location
Chr. 14: 38247388 - 38474931 on Build Rnor_6.0
UniGene ID
Rn.211679
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182473.1 NP_872279.1 14 2115
XM_008770139.1 XP_008768361.1 14 2162
GenBank mRNA AY251285.1 - 14 2115

View Full Product Details