Gene Transcripts

Gene Symbol
Lap3
Entrez Gene ID
289668
Gene Name
leucine aminopeptidase 3
Gene Aliases
LAP-3
Chromosome Location
Chr. 14: 70102813 - 70121797 on Build Rnor_6.0
UniGene ID
Rn.99790
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011910.1 NP_001011910.1 6 690
GenBank mRNA BC079381.1 - 6 690
FQ218842.1 - 2 208
FQ235301.1 - 6 699

View Full Product Details