Gene Transcripts

Gene Symbol
Morc2
Entrez Gene ID
289736
Gene Name
MORC family CW-type zinc finger 2
Gene Aliases
Morc2a, Zcwcc1
Chromosome Location
Chr. 14: 83889140 - 83930263 on Build Rnor_6.0
UniGene ID
Rn.224734
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106016.1 NP_001099486.1 20 2343
XM_006251198.2 XP_006251260.1 22 2486

View Full Product Details