Gene Transcripts

Gene Symbol
Mtfp1
Entrez Gene ID
289745
Gene Name
mitochondrial fission process 1
Gene Aliases
Mtp18
Chromosome Location
Chr. 14: 84330223 - 84334063 on Build Rnor_6.0
UniGene ID
Rn.106244
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006960.1 NP_001006961.1 4 1325
GenBank mRNA BC083713.1 - 4 1325

View Full Product Details