Gene Transcripts

Gene Symbol
Emc9
Entrez Gene ID
290224
Gene Name
ER membrane protein complex subunit 9
Gene Aliases
Fam158a, RGD1308113
Chromosome Location
Chr. 15: 34258822 - 34263156 on Build Rnor_6.0
UniGene ID
Rn.163298
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008296.1 NP_001008297.1 3, 4 490
GenBank mRNA BC086432.1 - 3, 4 490

View Full Product Details