Gene Transcripts

Gene Symbol
U2AF1
Entrez Gene ID
7307
Gene Name
U2 small nuclear RNA auxiliary factor 1
Gene Aliases
FP793, RN, RNU2AF1, U2AF35, U2AFBP
Chromosome Location
Chr. 21: 43092956 - 43108291 on Build GRCh38
UniGene ID
Hs.365116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025203.1 NP_001020374.1 2 187
NM_001025204.1 NP_001020375.1 2 187
NM_006758.2 NP_006749.1 2 187
XM_011529743.2 XP_011528045.1 2 900
XM_017028468.1 XP_016883957.1 2 900
GenBank mRNA AB451244.1 - 2 103
AF370386.1 - 1 910
AJ627978.1 - 2 133
AK307560.1 - 2 136
AL832665.1 - 2 133
BC001177.1 - 2 131 3534272
BC001923.1 - 2 131 2823296
BC005915.1 - 2 133 4295037
CB110007.1 - 2 187
M96982.1 - 2 141

Gene Symbol
U2AF1L5
Entrez Gene ID
102724594
Gene Name
U2 small nuclear RNA auxiliary factor 1 like 5
Gene Aliases
Chromosome Location
Chr. 21: 6484623 - 6499284 on Build GRCh38
UniGene ID
Hs.365116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320646.1 NP_001307575.1 2 215
NM_001320648.1 NP_001307577.1 2 215
NM_001320650.1 NP_001307579.1 2 215
NM_001320651.1 NP_001307580.1 2 215
XM_017028219.1 XP_016883708.1 2 215
XM_017028220.1 XP_016883709.1 2 215
GenBank mRNA AL522322.3 - 2 132
BG612658.1 - 2 180
CN280267.1 - 2 92
CN280269.1 - 2 152
DA211043.1 - 2 215

View Full Product Details