Gene Transcripts

Gene Symbol
Ly86
Entrez Gene ID
291359
Gene Name
lymphocyte antigen 86
Gene Aliases
Chromosome Location
Chr. 17: 28104535 - 28191453 on Build Rnor_6.0
UniGene ID
Rn.3176
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106128.1 NP_001099598.1 2 273
XM_006253800.2 XP_006253862.1 3 445

View Full Product Details