Gene Transcripts

Gene Symbol
Pabpc2
Entrez Gene ID
291615
Gene Name
poly(A) binding protein, cytoplasmic 2
Gene Aliases
Chromosome Location
Chr. 18: 32964763 - 32967031 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106151.2 NP_001099621.1 1 815

View Full Product Details