Gene Transcripts

Gene Symbol
Tepp
Entrez Gene ID
291850
Gene Name
testis, prostate and placenta expressed
Gene Aliases
Chromosome Location
Chr. 19: 10127607 - 10138436 on Build Rnor_6.0
UniGene ID
Rn.62971
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201655.1 NP_970617.1 4 592
XM_008772274.1 XP_008770496.1 4 869
XM_008772275.1 XP_008770497.1 4 869
XM_008772276.1 XP_008770498.1 4 869

View Full Product Details