Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039713.1 | NP_001034802.1 | 3 | 600 | |
XM_006255176.2 | XP_006255238.1 | 3 | 1033 | ||
XM_008772284.1 | XP_008770506.1 | 3 | 929 | ||
XM_008772285.1 | XP_008770507.1 | 3 | 931 | ||
GenBank mRNA | AM233906.1 | - | 3 | 600 | |
BC168239.1 | - | 3 | 615 | 5599736 | |
FQ211940.1 | - | 3 | 640 | ||
FQ212297.1 | - | 3 | 629 |