Gene Transcripts

Gene Symbol
Cog8
Entrez Gene ID
291990
Gene Name
component of oligomeric golgi complex 8
Gene Aliases
Chromosome Location
Chr. 19: 39246656 - 39257406 on Build Rnor_6.0
UniGene ID
Rn.3024
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106182.2 NP_001099652.1 3 1311
GenBank mRNA BC168966.1 - 3 1314

View Full Product Details