Gene Transcripts

Gene Symbol
Olr190
Entrez Gene ID
293334
Gene Name
olfactory receptor 190
Gene Aliases
ratchr1-162952790-162953737_ORF
Chromosome Location
Chr. 1: 169966190 - 169967137 on Build Rnor_6.0
UniGene ID
Rn.141819
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000185.1 NP_001000185.1 1 744

View Full Product Details