Gene Transcripts

Gene Symbol
Ndufs8
Entrez Gene ID
293652
Gene Name
NADH dehydrogenase (ubiquinone) Fe-S protein 8
Gene Aliases
Chromosome Location
Chr. 1: 219141289 - 219144610 on Build Rnor_6.0
UniGene ID
Rn.3373
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106322.2 NP_001099792.1 6 476
XM_006230731.1 XP_006230793.1 6 687
GenBank mRNA BC158791.1 - 6 476

View Full Product Details