Gene Transcripts

Gene Symbol
Olr343
Entrez Gene ID
293788
Gene Name
olfactory receptor 343
Gene Aliases
Chromosome Location
Chr. 1: 229803670 - 229804614 on Build Rnor_6.0
UniGene ID
Rn.141692
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000254.1 NP_001000254.1 1 838

View Full Product Details