Gene Transcripts

Gene Symbol
Olr376
Entrez Gene ID
293809
Gene Name
olfactory receptor 376
Gene Aliases
ratchr1-217038896-217039843_ORF
Chromosome Location
Chr. 1: 230628996 - 230629943 on Build Rnor_6.0
UniGene ID
Rn.141675
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000260.1 NP_001000260.1 1 272

View Full Product Details