Gene Transcripts

Gene Symbol
Olr385
Entrez Gene ID
294109
Gene Name
olfactory receptor 385
Gene Aliases
Olfr547, ratchr1-258329042-258330004_ORF
Chromosome Location
Chr. 1: 272859580 - 272892440 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760585.1 XP_008758807.1 2 825

View Full Product Details