Gene Transcripts
- Gene Symbol
- Cfb
- Entrez Gene ID
- 294257
- Gene Name
- complement factor B
- Gene Aliases
- Bf, Da1-24
- Chromosome Location
- Chr. 20: 4536206 - 4542073 on Build Rnor_6.0
- UniGene ID
- Rn.109148
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_212466.3 |
NP_997631.2
|
13 |
1884 |
|
GenBank mRNA |
BC087089.1 |
- |
13 |
1884 |
|
View Full Product Details