Gene Transcripts

Gene Symbol
Skiv2l
Entrez Gene ID
294260
Gene Name
superkiller viralicidic activity 2-like (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 20: 4519457 - 4530172 on Build Rnor_6.0
UniGene ID
Rn.138027
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213559.2 NP_998724.1 21 2548
XM_006255923.1 XP_006255985.1 21 2611

View Full Product Details