Gene Transcripts

Gene Symbol
Gprc6a
Entrez Gene ID
294394
Gene Name
G protein-coupled receptor, class C, group 6, member A
Gene Aliases
Chromosome Location
Chr. 20: 32607653 - 32628953 on Build Rnor_6.0
UniGene ID
Rn.140864
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271106.1 NP_001258035.1 6 2411
GenBank mRNA AJ535460.1 - 6 2387

View Full Product Details