Gene Transcripts

Gene Symbol
Slc50a1
Entrez Gene ID
295245
Gene Name
solute carrier family 50 (sugar efflux transporter), member 1
Gene Aliases
RGD1308478, Rag1ap1
Chromosome Location
Chr. 2: 188585118 - 188587498 on Build Rnor_6.0
UniGene ID
Rn.823
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106445.1 NP_001099915.1 2 240

View Full Product Details