Gene Transcripts

Gene Symbol
ZFY
Entrez Gene ID
7544
Gene Name
zinc finger protein, Y-linked
Gene Aliases
ZNF911
Chromosome Location
Chr. Y: 2934402 - 2982508 on Build GRCh38
UniGene ID
Hs.522845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145275.1 NP_001138747.1 3 957
NM_001145276.1 NP_001138748.1 3 556
NM_003411.3 NP_003402.2 4 1046
XM_005262570.2 XP_005262627.1 4 1129
XM_011531473.2 XP_011529775.1 3 2028
XM_011531474.2 XP_011529776.1 3 1705
XM_011531475.2 XP_011529777.1 4 2119
XM_017030075.1 XP_016885564.1 4 925
XM_017030076.1 XP_016885565.1 4 1129
XM_017030077.1 XP_016885566.1 3 1709
GenBank mRNA AK301061.1 - 3 238
AK310553.1 - 4 1129
BC114526.1 - 3 783 40054050
BC114960.1 - 4 872 40054055
L10393.1 - 3 758
M30607.1 - 3 725

View Full Product Details