Gene Transcripts

Gene Symbol
Dlx1
Entrez Gene ID
296500
Gene Name
distal-less homeobox 1
Gene Aliases
Chromosome Location
Chr. 3: 58164931 - 58169523 on Build Rnor_6.0
UniGene ID
Rn.59152
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100531.1 NP_001094001.1 2 952
GenBank mRNA BC167073.1 - 2 602

View Full Product Details