Gene Transcripts

Gene Symbol
H6pd
Entrez Gene ID
298655
Gene Name
hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase)
Gene Aliases
H6pdh
Chromosome Location
Chr. 5: 166998881 - 167030441 on Build Rnor_6.0
UniGene ID
Rn.17292
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106698.1 NP_001100168.1 2 793
XM_006239459.2 XP_006239521.1 2 943
XM_006239460.2 XP_006239522.1 3 1078

View Full Product Details