Gene Transcripts

Gene Symbol
Theg
Entrez Gene ID
299599
Gene Name
theg spermatid protein
Gene Aliases
Chromosome Location
Chr. 7: 13013521 - 13023666 on Build Rnor_6.0
UniGene ID
Rn.49140
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013102.1 NP_001013120.1 4 547
XM_006240857.2 XP_006240919.1 4 567
XM_006240858.2 XP_006240920.1 3 495
GenBank mRNA BC083921.1 - 4 547

View Full Product Details