Gene Transcripts

Gene Symbol
Lynx1
Entrez Gene ID
300018
Gene Name
Ly6/neurotoxin 1
Gene Aliases
Chromosome Location
Chr. 7: 115982879 - 115988082 on Build Rnor_6.0
UniGene ID
Rn.32045
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130546.1 NP_001124018.1 4 556
GenBank mRNA FQ212028.1 - 4 564

View Full Product Details