Gene Transcripts

Gene Symbol
Sh3bp1
Entrez Gene ID
300067
Gene Name
SH3-domain binding protein 1
Gene Aliases
Chromosome Location
Chr. 7: 120125714 - 120138148 on Build Rnor_6.0
UniGene ID
Rn.9374
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171981.1 NP_001165452.1 15 1381
XM_008765682.1 XP_008763904.1 14 2446

View Full Product Details