Gene Transcripts

Gene Symbol
Mbd3l1
Entrez Gene ID
300389
Gene Name
methyl-CpG binding domain protein 3-like 1
Gene Aliases
Chromosome Location
Chr. 8: 18428035 - 18434704 on Build Rnor_6.0
UniGene ID
Rn.124774
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106803.1 NP_001100273.1 3 360
XM_006242571.2 XP_006242633.1 2 445
XM_006242574.2 XP_006242636.1 3 721
XM_006242576.2 XP_006242638.1 2 276

View Full Product Details