Gene Transcripts

Gene Symbol
Plekhb2
Entrez Gene ID
301337
Gene Name
pleckstrin homology domain containing, family B (evectins) member 2
Gene Aliases
Chromosome Location
Chr. 9: 41348447 - 41381092 on Build Rnor_6.0
UniGene ID
Rn.1410
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106899.1 NP_001100369.1 6 461

View Full Product Details