Gene Transcripts

Gene Symbol
Rufy4
Entrez Gene ID
301510
Gene Name
RUN and FYVE domain containing 4
Gene Aliases
RGD1560678
Chromosome Location
Chr. 9: 81401071 - 81422011 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245288.2 XP_006245350.1 11 2128

View Full Product Details