Gene Transcripts

Gene Symbol
Cntnap5b
Entrez Gene ID
301650
Gene Name
contactin associated protein-like 5B
Gene Aliases
Caspr5-2, Caspr5-3, Cntnap5c, RGD1565194
Chromosome Location
Chr. 13: 21678088 - 22591058 on Build Rnor_6.0
UniGene ID
Rn.202933
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047873.1 NP_001041338.1 20 3237
XM_008769451.1 XP_008767673.1 20 5989
XM_008769452.1 XP_008767674.1 20 3670

View Full Product Details