Gene Transcripts

Gene Symbol
Ranbp17
Entrez Gene ID
303029
Gene Name
RAN binding protein 17
Gene Aliases
RGD1562576
Chromosome Location
Chr. 10: 18139763 - 18445623 on Build Rnor_6.0
UniGene ID
Rn.101744
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106994.1 NP_001100464.1 12 1388
XM_008767582.1 XP_008765804.1 12 3077
XM_008767583.1 XP_008765805.1 12 3050

View Full Product Details