Gene Transcripts
- Gene Symbol
- Ybx2
- Entrez Gene ID
- 303250
- Gene Name
- Y box binding protein 2
- Gene Aliases
- RGD1305068
- Chromosome Location
- Chr. 10: 56546689 - 56552338 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_220618.8 |
XP_220618.3
|
5 |
596 |
|
View Full Product Details