Gene Transcripts

Gene Symbol
Tm4sf5
Entrez Gene ID
303256
Gene Name
transmembrane 4 L six family member 5
Gene Aliases
Chromosome Location
Chr. 10: 57111907 - 57118299 on Build Rnor_6.0
UniGene ID
Rn.153640
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107015.1 NP_001100485.1 4 562

View Full Product Details