Gene Transcripts

Gene Symbol
Cpox
Entrez Gene ID
304024
Gene Name
coproporphyrinogen oxidase
Gene Aliases
Chromosome Location
Chr. 11: 44039665 - 44049648 on Build Rnor_6.0
UniGene ID
Rn.19581
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037095.1 NP_001032172.1 6 1284
GenBank mRNA BC107664.1 - 6 1284

View Full Product Details