Gene Transcripts

Gene Symbol
Epb4.1l5
Entrez Gene ID
304733
Gene Name
erythrocyte membrane protein band 4.1-like 5
Gene Aliases
Epb41l5
Chromosome Location
Chr. 13: 35622472 - 35673449 on Build Rnor_6.0
UniGene ID
Rn.2480
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012023.1 NP_001012023.1 12 1027
GenBank mRNA BC090012.1 - 12 1027

View Full Product Details