Gene Transcripts

Gene Symbol
Zbtb37
Entrez Gene ID
304918
Gene Name
zinc finger and BTB domain containing 37
Gene Aliases
RGD1308731
Chromosome Location
Chr. 13: 78836491 - 78852211 on Build Rnor_6.0
UniGene ID
Rn.218569
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107191.1 NP_001100661.1 5 1606
XM_006250110.2 XP_006250172.1 4 1461
XM_006250111.2 XP_006250173.1 4 1464
XM_008769648.1 XP_008767870.1 5 1654

View Full Product Details