Gene Transcripts

Gene Symbol
Atf6
Entrez Gene ID
304962
Gene Name
activating transcription factor 6
Gene Aliases
Chromosome Location
Chr. 13: 89053457 - 89242528 on Build Rnor_6.0
UniGene ID
Rn.15235
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107196.1 NP_001100666.1 7, 8 939
XM_008769738.1 XP_008767960.1 7, 8 995
GenBank mRNA BC168890.1 - 7, 8 922

View Full Product Details