Gene Transcripts

Gene Symbol
Dok7
Entrez Gene ID
305448
Gene Name
docking protein 7
Gene Aliases
RGD1566416
Chromosome Location
Chr. 14: 80926071 - 80959398 on Build Rnor_6.0
UniGene ID
Rn.19295
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130062.1 NP_001123534.1 2 71
XM_006251239.2 XP_006251301.1 3 888
XM_008770283.1 XP_008768505.1 3 888
GenBank mRNA FQ223777.1 - 3 190

View Full Product Details