Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008770390.1 | XP_008768612.1 | 10 | 1264 | |
XM_008770391.1 | XP_008768613.1 | 10 | 1264 | ||
XM_008770392.1 | XP_008768614.1 | 9 | 1090 | ||
XM_008770393.1 | XP_008768615.1 | 5 | 2006 | ||
GenBank mRNA | AB095021.1 | - | 10 | 1133 | |
BC083629.1 | - | 10 | 1238 | 7134382 |