Gene Transcripts

Gene Symbol
LOC305806
Entrez Gene ID
305806
Gene Name
similar to glutaredoxin 1 (thioltransferase); glutaredoxin
Gene Aliases
Chromosome Location
Chr. 15: 5780834 - 5794911 on Build Rnor_6.0
UniGene ID
Rn.123447
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013993.1 NP_001014015.1 512
GenBank mRNA BC079250.1 - 512

View Full Product Details