Gene Transcripts

Gene Symbol
Hspa14
Entrez Gene ID
307133
Gene Name
heat shock protein 14
Gene Aliases
Chromosome Location
Chr. 17: 78735630 - 78756412 on Build Rnor_6.0
UniGene ID
Rn.137392
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004257.1 NP_001004257.1 3 249
GenBank mRNA BC079118.1 - 3 249

View Full Product Details