Gene Transcripts

Gene Symbol
Atxn1l
Entrez Gene ID
307970
Gene Name
ataxin 1-like
Gene Aliases
RGD1561758
Chromosome Location
Chr. 19: 41932356 - 41939456 on Build Rnor_6.0
UniGene ID
Rn.23514
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772565.1 XP_008770787.1 1 720

View Full Product Details