Gene Transcripts

Gene Symbol
HIST1H2BF
Entrez Gene ID
8343
Gene Name
histone cluster 1, H2bf
Gene Aliases
H2B/g, H2BFG
Chromosome Location
Chr. 6: 26199559 - 26199988 on Build GRCh38
UniGene ID
Hs.182137
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003522.3 NP_003513.1 1 87
GenBank mRNA BC096120.1 - 1 87
BC096123.1 - 1 87 40002357

View Full Product Details